Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059708.4 | NP_492109.2 | 14 - 15 | 1704 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |