Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcp-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059710.7 | NP_492111.3 | 4 - 5 | 1644 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |