Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306353.3 | NP_001293282.1 | 5 - 6 | 972 | 147 | |
NM_059723.7 | NP_492124.1 | 7 - 8 | 1210 | 147 | ||
NM_059724.4 | NP_492125.1 | 6 - 7 | 1228 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |