Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263616.4 | NP_001250545.1 | 8 - 9 | 1398 | 82 | |
NM_001306353.3 | NP_001293282.1 | 6 - 7 | 1182 | 82 | ||
NM_001306354.3 | NP_001293283.1 | 6 - 7 | 1158 | 82 | ||
NM_059723.7 | NP_492124.1 | 8 - 9 | 1420 | 82 | ||
NM_059724.4 | NP_492125.1 | 7 - 8 | 1438 | 82 |
Gene Symbol: D2030.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059722.6 | NP_492123.2 | 17 - 18 | 4747 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |