Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263626.3 | NP_001250555.1 | 13 - 14 | 3173 | 70 | |
NM_001263627.3 | NP_001250556.1 | 12 - 13 | 2920 | 70 | ||
GenBank mRNA | AF159144.1 | - | 13 - 14 | 3173 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |