Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263626.3 | NP_001250555.1 | 17 - 18 | 4291 | 154 | |
NM_001263627.3 | NP_001250556.1 | 16 - 17 | 4038 | 154 | ||
GenBank mRNA | AF159144.1 | - | 17 - 18 | 4291 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |