Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026460.3 | NP_001021631.1 | 5 - 6 | 1367 | 89 | |
NM_001026461.4 | NP_001021632.1 | 5 - 6 | 1365 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |