Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mau-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059827.6 | NP_492228.1 | 7 - 8 | 1305 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |