Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mau-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059827.6 | NP_492228.1 | 8 - 9 | 1730 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |