Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kri-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026692.5 | NP_001021863.1 | 2 - 3 | 133 | 81 | |
NM_001026693.5 | NP_001021864.1 | 2 - 3 | 128 | 81 | ||
GenBank mRNA | DQ372920.1 | - | 2 - 3 | 128 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |