Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026307.7 | NP_001021478.1 | 9 - 10 | 1351 | 137 | |
NM_001383463.1 | 8 - 9 | 1025 | 137 | |||
GenBank mRNA | AF440800.2 | - | 8 - 9 | 1031 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |