Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ngp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383469.1 | 4 - 5 | 426 | 107 | ||
NM_059874.6 | NP_492275.2 | 4 - 5 | 451 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |