Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ngp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383469.1 | 6 - 7 | 727 | 97 | ||
NM_059874.6 | NP_492275.2 | 6 - 7 | 752 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |