Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128970.4 | NP_001122442.1 | 7 - 8 | 2096 | 83 | |
NM_059883.8 | NP_492284.2 | 7 - 8 | 2097 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |