Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059903.6 | NP_492304.1 | 3 - 4 | 1068 | 121 | |
GenBank mRNA | AF286377.1 | - | 3 - 4 | 807 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |