Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zhp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263872.2 | NP_001250801.1 | 5 - 6 | 803 | 119 | |
NM_001263873.4 | NP_001250802.1 | 5 - 6 | 815 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |