Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sig-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373191.2 | NP_001359651.1 | 4 - 5 | 547 | 70 | |
NM_059942.8 | NP_492343.2 | 4 - 5 | 683 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |