Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hot-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059948.6 | NP_492349.1 | 1 - 2 | 185 | 90 | |
GenBank mRNA | AF324057.1 | - | 1 - 2 | 58 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |