Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129002.4 | NP_001122474.1 | 8 - 9 | 1101 | 83 | |
NM_059955.7 | NP_492356.1 | 8 - 9 | 1090 | 83 | ||
GenBank mRNA | AF228692.1 | - | 8 - 9 | 1112 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |