Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373211.3 | NP_001359628.1 | 9 - 10 | 1562 | 75 | |
NM_059972.6 | NP_492373.1 | 10 - 11 | 1646 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |