Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cids-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128999.4 | NP_001122471.1 | 1 - 2 | 101 | 64 | |
NM_059979.6 | NP_492380.2 | 1 - 2 | 110 | 64 | ||
NM_182083.6 | NP_871883.2 | 1 - 2 | 101 | 64 | ||
NR_023944.1 | 1 - 2 | 37 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |