Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thoc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060015.5 | NP_492416.2 | 8 - 9 | 1075 | 127 |
Gene Symbol: F32H2.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060014.5 | NP_492415.1 | 7 - 8 | 1196 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |