Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136284.4 | NP_001129756.1 | 8 - 9 | 1387 | 103 | |
NM_060028.7 | NP_492429.2 | 8 - 9 | 1388 | 103 | ||
GenBank mRNA | AY523514.1 | - | 8 - 9 | 1388 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |