Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060032.7 | NP_492433.1 | 6 - 7 | 1005 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |