Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad-54.L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060037.5 | NP_492438.1 | 10 - 11 | 1482 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |