Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026506.5 | NP_001021677.3 | 2 - 3 | 160 | 68 | |
NM_001026507.5 | NP_001021678.3 | 2 - 3 | 412 | 68 | ||
NM_001026508.6 | NP_001021679.3 | 2 - 3 | 161 | 68 | ||
NM_060041.6 | NP_492442.3 | 2 - 3 | 146 | 68 | ||
NM_060042.5 | NP_492443.3 | 2 - 3 | 138 | 68 | ||
NM_060043.5 | NP_492444.4 | 2 - 3 | 146 | 68 | ||
NR_000051.1 | 2 - 3 | 103 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |