Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276534.2 | NP_001263463.1 | 3 - 4 | 723 | 148 | |
NM_060048.11 | NP_492449.2 | 4 - 5 | 871 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |