Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026238.5 | NP_001021409.2 | 4 - 5 | 643 | 120 | |
NM_001030057.4 | NP_001025228.2 | 6 - 7 | 1223 | 120 | ||
NM_001313465.3 | NP_001300394.1 | 11 | 1389 | 120 | ||
NM_001313466.5 | NP_001300395.1 | 12 | 1479 | 120 | ||
NM_001377616.3 | NP_001364651.1 | 9 - 10 | 1068 | 120 | ||
NM_001377617.3 | NP_001364650.1 | 6 - 7 | 792 | 120 | ||
GenBank mRNA | EF443133.1 | - | 9 - 10 | 1068 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |