Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030057.4 | NP_001025228.2 | 1 - 2 | 576 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |