Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313465.3 | NP_001300394.1 | 1 | 100 | 136 | |
NM_001313466.5 | NP_001300395.1 | 2 | 190 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |