Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C45G3.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381008.1 | NP_001366919.1 | 12 - 13 | 2840 | 140 |
Gene Symbol: gip-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025873.6 | NP_001021044.1 | 8 - 9 | 1775 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |