Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tat-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026286.5 | NP_001021457.1 | 4 - 5 | 1190 | 146 | |
NM_001026287.6 | NP_001021458.2 | 5 - 6 | 1097 | 146 | ||
NM_001047200.3 | NP_001040665.1 | 4 - 5 | 1181 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |