Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025827.5 | NP_001020998.1 | 7 - 8 | 1333 | 73 | |
NM_001025828.6 | NP_001020999.1 | 7 - 8 | 1333 | 73 | ||
GenBank mRNA | AF195610.1 | - | 6 - 7 | 1138 | 73 | |
AF195611.1 | - | 6 - 7 | 1138 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |