Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026296.3 | NP_001021467.1 | 3 - 4 | 1572 | 74 | |
NM_001026297.6 | NP_001021468.1 | 3 - 4 | 447 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |