Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381092.1 | NP_001366862.1 | 4 - 5 | 378 | 113 | |
NM_060134.4 | NP_492535.1 | 4 - 5 | 568 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |