Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128955.4 | NP_001122427.1 | 6 - 7 | 851 | 76 | |
NM_001128956.4 | NP_001122428.1 | 1 - 2 | 71 | 76 | ||
NM_001264200.4 | NP_001251129.1 | 6 - 7 | 851 | 76 | ||
NM_001264202.1 | NP_001251131.1 | 6 - 7 | 851 | 76 | ||
NM_001264203.2 | NP_001251132.1 | 6 - 7 | 698 | 76 | ||
NM_001381092.1 | NP_001366862.1 | 5 - 6 | 499 | 76 | ||
NM_060134.4 | NP_492535.1 | 5 - 6 | 689 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |