Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060167.7 | NP_492568.1 | 4 - 5 | 2072 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |