Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047198.5 | NP_001040663.1 | 6 - 7 | 1125 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |