Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264305.3 | NP_001251234.1 | 3 - 4 | 901 | 79 | |
NM_001381165.1 | NP_001367039.1 | 6 - 7 | 1342 | 79 | ||
NM_001381168.2 | NP_001366915.1 | 6 - 7 | 1428 | 79 | ||
NM_060212.7 | NP_492613.1 | 6 - 7 | 1406 | 79 | ||
NM_181999.6 | NP_871799.1 | 6 - 7 | 1297 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |