Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060214.5 | NP_492615.2 | 3 - 4 | 374 | 110 | |
GenBank mRNA | AY204178.1 | - | 3 - 4 | 394 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |