Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oct-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026311.7 | NP_001021482.1 | 9 - 10 | 1083 | 191 | |
NM_060216.6 | NP_492617.4 | 10 - 11 | 1146 | 191 | ||
GenBank mRNA | AF110415.1 | - | 9 - 10 | 1091 | 191 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |