Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060219.9 | NP_492620.1 | 9 - 10 | 1568 | 159 | |
GenBank mRNA | AF143242.1 | - | 9 - 10 | 1559 | 159 | |
AF145377.1 | - | 9 - 10 | 1350 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |