Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026468.8 | NP_001021639.3 | 2 - 3 | 1142 | 89 | |
NM_001026469.8 | NP_001021640.2 | 3 - 4 | 1562 | 89 | ||
NM_001313510.3 | NP_001300439.1 | 3 - 4 | 1568 | 89 | ||
GenBank mRNA | AB088397.1 | - | 2 - 3 | 1142 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |