Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313508.2 | NP_001300437.1 | 2 - 3 | 911 | 82 | |
NM_060234.7 | NP_492635.1 | 2 - 3 | 911 | 82 | ||
GenBank mRNA | AF013953.1 | - | 2 - 3 | 903 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |