Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sys-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313509.3 | NP_001300438.1 | 2 - 3 | 279 | 146 | |
NM_001383706.1 | 4 - 5 | 689 | 146 | |||
NM_060238.6 | NP_492639.1 | 4 - 5 | 686 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |