Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fog-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060245.6 | NP_492646.1 | 6 - 7 | 584 | 88 | |
GenBank mRNA | AJ011777.1 | - | 6 - 7 | 599 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |