Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060295.5 | NP_492696.1 | 6 - 7 | 868 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |