Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026294.3 | NP_001021465.1 | 1 - 2 | 158 | 62 | |
NM_001373640.1 | NP_001359696.1 | 1 - 2 | 158 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |