Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbtr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129070.2 | NP_001122542.1 | 3 - 4 | 555 | 78 | |
NM_001129071.3 | NP_001122543.1 | 3 - 4 | 493 | 78 | ||
GenBank mRNA | DQ904352.1 | - | 3 - 4 | 495 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |