Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306301.3 | NP_001293230.1 | 7 - 8 | 1465 | 78 | |
NM_001306302.3 | NP_001293231.1 | 1 - 2 | 115 | 78 | ||
NM_058427.6 | NP_490828.3 | 9 - 10 | 2033 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |